missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ Turbo 15 PES Centrifugal Concentrators
BIOPHARMA

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

Brand:  Sartorius VS15T92

513.99 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 12654863

  • £773.00 / Pack of 48
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

Twin vertical membranes and a sleek internal design for ultra-fast concentration
  • In a single spin, 15 ml solutions can be concentrated up to 150x in 10-30 minutes
  • Large, high-flux PES membranes feature ultra-low protein binding and low extractables
  • High, constant filtration pressure allows optimal crossflow to minimize membrane fouling
Designed for maximum convenience, safety and sample recovery
  • Angular, pipette-friendly pocket facilitates easy retentate recovery of greater than 95%
  • Eliminates the risk of sample filtration to dryness
  • Stable, inert polystyrene housing with high chemical compatibility (pH 1-14)
  • UV joining technology for a smooth transition between membrane and housing
  • Volume graduation for easy monitoring of concentrate volume and 100μl dead-stop volume
Universal compatibility for rotors that accept 50mL conical bottom tubes
  • Handles sample volumes up to 15mL in swing bucket rotors (11mL in fixed angle rotors)
  • Designed for rotors that accommodate Falcon 50mL conical tubes:
    • Beckman Allegra 25R with TS-5.1-500 swing-out rotor, BUC 5 buckets, adaptors
    • Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
    • Multifuge 3 S-R with 75006445 rotor, 75006441 buckets, adaptors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Centrifuge
3 kDa
Non-sterile
Concentration, desalting or buffer exchange applications
4,000 x g
Centrifugal Concentrator
11.8 cm
Ultrafiltration
PES
15 mL
7.2 cm2
Polystyrene
48/Pk.
11 mL
27 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number240300157 Certificate TypeCertificate of Quality Date01/12/2024 Catalog NumberVS15T92
Lot Number240100817 Certificate TypeCertificate of Quality Date12/10/2024 Catalog NumberVS15T92
Lot Number234500107 Certificate TypeCertificate of Quality Date10/08/2024 Catalog NumberVS15T92
Lot Number232200587 Certificate TypeCertificate of Quality Date03/02/2024 Catalog NumberVS15T92
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Vivaspin™ Turbo 15 PES Centrifugal Concentrators > 3,000 MWCO; 48/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.