missing translation for 'onlineSavingsMsg'
Learn More
Scandium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11336268

Scandium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11336268
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11336268

Brand: Thermo Scientific Alfa Aesar 011219.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20662-14-0
Molecular Formula Cl3H12O6Sc
Molecular Weight (g/mol) 259.40
MDL Number MFCD00149852
InChI Key FKZFOHABAHJDIK-UHFFFAOYSA-K
IUPAC Name scandium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Sc+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 1 g
Merck Index 14,8392
Formula Weight 259.41 (151.32 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Scandium(III) chloride hexahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ27K039 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number011219.06,011219.03
Lot NumberZ27K045 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number011219.06,011219.03
Lot NumberP22K066 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog Number011219.06,011219.03
Lot NumberX26D004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011219.06,011219.03
Lot NumberF12L21 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011219.06,011219.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scandium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.