Learn More
3M™ SecureFit™ SF600 Series Safety Glasses
Comprised of a semi-rimless lens and fixed-length temple arm. 3M™ SecureFit™ SF600 Series Safety Glasses include an integrated side shield for added protection.
£14.79 - £68.70
Specifications
Frame Color Gray
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection Yes
View More Specs
Products 9
Product Code Brand Lens Coating Lens Type Lens Tint Price Quantity & Availability  
Product Code Brand Lens Coating Lens Type Lens Tint Price Quantity & Availability  
15727727
View Documents
3M™
7100112718
Scratch-resistant I/O Mirror £14.79
Each
Qty Price Break Available
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
15747727
View Documents Promotion Details
3M™
7100112721
Scratch-resistant Welding shade 1.7
£25.25
Each
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
15757727
View Documents Promotion Details
3M™
7100112724
Scratch-resistant Welding shade 3.0
£25.25
Each
In Stock
to see stock.
Edge
Add to basket
15797717
View Documents Promotion Details
3M™
7100112715
Anti-fog, Scratch-resistant Amber
£27.30
Each
In Stock
to see stock.
Edge
Add to basket
15737727
View Documents
3M™
7100112720
Scratch-resistant Clear
£16.20
Each
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
15787717
View Documents
3M™
7100112713
Scratch-resistant Polarized Gray
£68.70
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
15707727
View Documents Promotion Details
3M™
7100112716
Anti-fog, Scratch-resistant Gray
£19.20
Each
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
15717727
View Documents Promotion Details
3M™
7100112717
Anti-fog, Scratch-resistant Clear
£19.20
Each
In Stock
to see stock.
Edge
Add to basket
15777717
View Documents
3M™
7100112712
Scratch-resistant Welding shade 5.0
£19.20
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Optical class 1 lens suitable for prolonged use
  • Design provides coverage and a good field of vision based on two lenses in a semi-rimless design
  • Offers protection against Ultraviolet (UV) radiation in accordance with European standards
  • Weight: 27g
  • 3M™ Pressure Diffusion Temple Technology for secure and comfortable fit
  • Soft nose pads for comfortable fit
  • Superior Scotchgard™ anti-fog coating meeting K and N on the standard clear, gray, and amber lenses
Specifications
Specifications
Gray
Polycarbonate
Polycarbonate
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
3M™ SecureFit™ SF600 Series Safety Glasses