Learn More
Cytiva SpeedBeads™ Magnetic Neutravidin Coated Particles, 1 um, 1% solids, 0.05% Na azide
Capture of Biotinylated Probes. Their cauliflower-like structure gives Neutravidin beads high binding capacity. Neutravidin SpeedBeads combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in immunoassay and molecular biology applications.
Brand:  Cytiva 78152104010150
723.90 GBP valid until 2025-12-31
Use promo code "24338" to get your promotional price.
 View more versions of this product
Product Code. 11528692
  • £762.00 / Each
Explore more special offers
Description
Description
  • Neutravidin beads exhibit high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond quickly to a magnetic field to shorten assay times, improve precision and move faster through viscous solutions
  • Available with a 3500-4500 pmol/mg biotin-binding range
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1% solids (10mg/mL)
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
5 mL
2°C to 8°C
0.05% Na azide
1 μm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva SpeedBeads™ Magnetic Neutravidin Coated Particles, 1 um, 1% solids, 0.05% Na azide > 5mL