missing translation for 'onlineSavingsMsg'
Learn More

SGE™ eVol™ Handheld Automated Analytical Syringe

Improves laboratory workflow

Brand:  SGE™ 2910324

 View more versions of this product

Product Code. 12957596

  • £265.00 / Pack of 3

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.

  • Digitally controlled positive displacement dispensing system
  • Programmable—enables user-independent liquid handling procedures
  • Easily calibrated to ensure accurate dispensing
  • Stores calibration factors for up to ten XCHANGE™ syringes
  • Touch Wheel user interface
  • Full color screen

Applications:

  • Preparation of calibration standards
  • Addition of internal standards
  • Precise dispensing of non-aqueous liquids
  • Sample dilution

Specifications

Specifications

25 G
0.32 mm
0.63 mm
eVol Analytical Syringe
500 μL
Bevel
0.012 in.
0.024 in.
1.96 in.
3/Pk.
50 mm
eVol Automated Syringe
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SGE™ eVol™ Handheld Automated Analytical Syringe > Electronic syringe; 500μL; 0.32mm I.D.; Bevel tip

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.