missing translation for 'onlineSavingsMsg'
Learn More
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™ Product Code.: 10621573

Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™

Product Code. 10621573
5 kg, Plastic Keg
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Keg
Plastic Powder Jar
This item is not returnable. View return policy

Product Code. 10621573

Brand: Fisher Chemical S/0761/65

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color Brown-Orange
Physical Form Solid
pH 1.5 to 5
Quantity 5 kg
Packaging Plastic Keg
IUPAC Name dioxosilane
Formula Weight 60.08g/mol
  • Fisher Chemical - Silica gel, self indicating, 4 to 7 mesh (2.5mm to 6.0mm)
    Silica gel, self indicating, 4-7 mesh (2.5mm to 6.0mm) colour change from orange to colourless as water is absorbed
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisher Chemical Silica gel, self indicating, 4 to 7 mesh (2.5mm to 6.0mm)

    • Multiple Manufacturers
      Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
      Promo Code: 24155

      Expires2025-04-30

      Hot Deal

      *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

      Terms & Conditions See Full Promo Details
    • Multiple Manufacturers
      Shop now and save up to 97% on top-quality products for your lab needs.
      Promo Code: 25908

      Expires2025-03-31

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™ >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.