Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Glass Plates
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on glass plates come in a variety of plate sizes and layer thicknesses. Select a fluorescent indicator and choose from a variety of pack sizes.
£75.66 - £393.00
Specifications
Material | Silica 60 |
---|---|
Particle Size | 5 to 17 μm |
Pore Size | 60 Å |
Type | Glass Plate |
Pore Volume | 0.75 mL/g |
Product Code | Brand | Dimensions (L x W) | Quantity | Thickness (Metric) | Fluorescent Indicator | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Quantity | Thickness (Metric) | Fluorescent Indicator | Price | Quantity & Availability | |||
10308221
|
Macherey-Nagel™
809020 |
100 x 100 mm | 25 Pk. | 0.25 mm | UV254 |
£75.66
Pack of 25 |
In Stock
Log in to see stock. |
|||
10469941
|
Macherey-Nagel™
809012 |
100 x 200 mm | 50 Pk. | 0.25 mm |
£184.00
Pack of 50 |
In Stock
Log in to see stock. |
||||
10649391
|
Macherey-Nagel™
809122 |
100 x 200 mm | 50 Pk. | 0.25 mm | UV254 + UV366 |
£184.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
11857043
|
Macherey-Nagel™
809022 |
100 x 200 mm | 50 Pk. | 0.25 mm | UV254 |
£393.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
10371102
|
Macherey-Nagel™
809023 |
200 x 200 mm | 25 Pk. | 0.25 mm | UV254 |
£355.00
Pack of 25 |
In Stock
Log in to see stock. ![]() |
|||
10015681
|
Macherey-Nagel™
809013 |
200 x 200 mm | 25/Pk. | 0.25 mm |
£356.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10023021
|
Macherey-Nagel™
809083 |
200 x 200 mm | 12 Pk. | 2 mm | UV254 |
£257.00
Pack of 12 |
In Stock
Log in to see stock. |
|||
10055711
|
Macherey-Nagel™
809123 |
200 x 200 cm | 25 Pk. | 0.25 mm | UV254 + UV366 |
£160.00
Pack of 25 |
In Stock
Log in to see stock. |
|||
10698811
|
Macherey-Nagel™
809051 |
200 x 200 mm | 20 Pk. | 0.5 mm |
£217.00
Pack of 20 |
In Stock
Log in to see stock. |
||||
10609011
|
Macherey-Nagel™
809061 |
200 x 200 mm | 15 Pk. | 1 mm |
£224.00
Pack of 15 |
In Stock
Log in to see stock. |
||||
10184851
|
Macherey-Nagel™
809073 |
200 x 200 mm | 12 Pk. | 2 mm |
£253.76
Pack of 12 |
In Stock
Log in to see stock. |
||||
10576701
|
Macherey-Nagel™
809017 |
50 x 100 mm | 50 Pk. | 0.25 mm |
£199.00
Pack of 50 |
In Stock
Log in to see stock. |
||||
10575571
|
Macherey-Nagel™
809027 |
50 x 100 mm | 50 Pk. | 0.25 mm | UV254 |
£245.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
10388071
|
Macherey-Nagel™
809011 |
50x200mm | 100 Pk. | 0.25 mm |
£214.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10716152
|
Macherey-Nagel™
809021 |
50 x 200 mm | 100 Pk. | 0.25 mm | UV254 |
£280.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
12770154
|
Macherey-Nagel™
809121 |
50 x 200 mm | 100 Pk. | 0.25 mm | UV254 + UV366 |
£244.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
Description
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on glass plates come in a variety of plate sizes and layer thicknesses. Select a fluorescent indicator and choose from a variety of pack sizes.
Glass TLC plates, SIL G/UV 254.- Pre-coated ready to use plates
- Broad range of plate sizes, surface coatings and backings
- Silica 60
- specific surface (BET) ∼ 500 m2/g
- Mean pore size 60 Å
- Specific pore volume 0.75 mL/g,
- Particle size 5–17 μm
- Standard grade
- For analytical plates 0.25 mm
- For preparative plates 0.5 and 1 mm
- For 2 mm preparative layers a slightly coarser material is used.
- Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
- Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
- Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications
Silica 60 | |
60 Å | |
0.75 mL/g |
5 to 17 μm | |
Glass Plate | |
∼500 m2/g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.