Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Glass Plates
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on glass plates come in a variety of plate sizes and layer thicknesses. Select a fluorescent indicator and choose from a variety of pack sizes.
Brand:  Macherey-Nagel™ 809027
153.12 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10575571
  • £245.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Show More
Specifications
Specifications
Silica 60
60 Å
Glass Plate
0.75 mL/g
0.25 mm
5 to 17 μm
50 Pk.
UV254
∼500 m2/g
50 x 100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Standard SIL G Silica Layers on Glass Plates > Size: 50x100mm