missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC34A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA578333
This item is not returnable.
View return policy
Description
Positive Control: Unboiled HeLa Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SLC34A2 encodes a protein that is a pH-sensitive sodium-dependent phosphate transporter. Phosphate uptake is increased at lower pH. Defects in this gene are a cause of pulmonary alveolar microlithiasis. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
SLC34A2 | |
Polyclonal | |
Unconjugated | |
SLC34A2 | |
AA536683; D5Ertd227e; Na(+)/Pi cotransporter 2B; Na(+)-dependent phosphate cotransporter 2B; NAPI IIb; NaPi-2b; NaPi3b; NAPI-3B; NAPI-IIb; Npt2b; NPTIIb; rNaPi IIb; Slc34a2; Sodium/phosphate cotransporter 2B; Sodium-dependent phosphate transport protein 2B; sodium-phosphate transport protein 2B; solute carrier family 34 (sodium phosphate), member 2; solute carrier family 34 (type II sodium/phosphate contransporter), member 2; solute carrier family 34 (type II sodium/phosphate cotransporter), member 2; solute carrier family 34 member 2; type II sodium-dependent phosphate transporter 3b; type IIb Na/Picotransporter; type IIb sodium-phosphate transporter | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10568, 84395 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
2.03 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O95436, Q9JJ09 | |
SLC34A2 | |
Recombinant protein encompassing a sequence within the C-terminus region of human SLC34A2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 14
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SLC34A2 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction