Learn More
Brannan™ Specific Gravity Hydrometers M100 Series
Shot loaded specific gravity hydrometers for a variety of surface tension ranges.
Brand: Brannan™ 10/947/0
Description
Compares the relative density of a given liquid with that of water at 15.6°C (60°F). Brannan™ M100 Series Specific Gravity Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
- Includes protective plastic tube
- 250 mm in length
- Scale subdivision 0.002
- Accuracy of 0.002 specific gravity
- Available adjusted for either low or medium surface tension
- Available for numerous specific gravity ranges
Specifications
1 Ea. | |
0.002 g/mL | |
ISO 650, 916 | |
Steel Shot | |
1.30 to 1.40 g/mL |
Specific Gravity Hydrometer | |
0.002 g/mL | |
250 mm | |
Relative density of a given liquid with that of water |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.