missing translation for 'onlineSavingsMsg'
Learn More
Sodium cyclamate, 98%, Thermo Scientific Chemicals Product Code.: 11412958

Sodium cyclamate, 98%, Thermo Scientific Chemicals

Product Code. 11412958
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11412958

Brand: Thermo Scientific Alfa Aesar A18666.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose's. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose′s. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.

Solubility
Soluble in water (>=10 g/100 mL at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-05-9
Molecular Formula C6H12NNaO3S
Molecular Weight (g/mol) 201.22
MDL Number MFCD00003827
InChI Key UDIPTWFVPPPURJ-UHFFFAOYSA-M
Synonym sodium cyclamate, sodium cyclohexylsulfamate, sodium n-cyclohexylsulfamate, cyclamate sodium, cyclamic acid sodium salt, assugrin, ibiosuc, suessette, suestamin, sugarinShow MoreShow Less
PubChem CID 23665706
ChEBI CHEBI:82431
SMILES [Na+].[O-]S(=O)(=O)NC1CCCCC1

Specifications

Melting Point >300°C
Quantity 250 g
Beilstein 4166868
Solubility Information Soluble in water (>=10g/100 mL at 20°C).
IUPAC Name sodium;N-cyclohexylsulfamate
Formula Weight 201.22
Percent Purity 98%
Chemical Name or Material Sodium cyclamate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10239898 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA18666.30,A18666.0I
Lot Number10161887 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18666.30,A18666.0I
Lot Number10186981 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18666.30,A18666.0I
Lot Number10195053 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18666.30,A18666.0I
Lot NumberE3471B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18666.30,A18666.0I
compliance-icons
Product Identifier
  • Sodium cyclamate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sodium cyclamate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium cyclamate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.