missing translation for 'onlineSavingsMsg'
Learn More
Sodium ethoxide, 96%, pure, Thermo Scientific Chemicals Product Code.: 10225600

Sodium ethoxide, 96%, pure, Thermo Scientific Chemicals

Product Code. 10225600
500 g, Glass bottle
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10225600

Brand: Thermo Scientific Acros 168595000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-52-6
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.04
MDL Number MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1Show MoreShow Less
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]

Specifications

Melting Point 260.0°C
Density 0.868
Assay Percent Range 95% min. (NaOC2H5)
Packaging Glass bottle
Linear Formula C2H5ONa
Quantity 500 g
Beilstein 01, 311
Fieser 01,1065; 03,265; 04,451; 06,540
Merck Index 15, 8749
Solubility Information Solubility in water: reacts. Other solubilities: soluble in absolute alcohol without decomposition
Formula Weight 68.04
Percent Purity 96%
Grade Pure
Chemical Name or Material Sodium ethoxide, 96%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461024 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number168595000,168590025,168592500
Lot NumberA0464923 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog Number168595000,168590025,168592500
Lot NumberA0461024 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number168595000,168590025,168592500
Lot NumberA0449910 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number168595000,168590025,168592500
Lot NumberA0456515 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number168595000,168590025,168592500
compliance-icons compliance-icons
Product Identifier
  • Sodium ethoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H251-Self-heating: may catch fire.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium ethoxide, 96%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.