Learn More
Sodium Nitrite 0.5 M (0.5 N) Analytical Volumetric Solution (AVL), Reagecon
Fully factorized, high purity, stable product, developed and tested for titrations.
£46.35 - £87.35
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11572992
|
Reagecon
NANO051 |
1 L | HDPE Bottle GL45 |
£46.35
1 L |
Estimated Shipment: 15-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11562992
|
Reagecon
NANO055 |
5 L | HDPE Container |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.