missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals Product Code.: 10168282

Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10168282
25 kg, Fibre drum
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10168282

Brand: Thermo Scientific Acros 196640251

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1Show MoreShow Less
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point >1700.0°C
Melting Point 884.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 kg
Assay Percent Range 98.5% min
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15, 8809
Solubility Information Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
IUPAC Name disodium sulfate
Formula Weight 142.04
Percent Purity 99%
Grade Extra Pure
Packaging Fibre drum
Density 2.68
Chemical Name or Material Sodium sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464822 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number196640050,196640025,196640250,196640010,196640251
Lot NumberA0464226 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number196640050,196640025,196640250,196640010,196640251
Lot NumberA0463652 Certificate TypeCertificate of Analysis Date21/06/2024 Catalog Number196640050,196640025,196640250,196640010,196640251
Lot NumberA0463781 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number196640050,196640025,196640250,196640010,196640251
Lot NumberA0463651 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog Number196640050,196640025,196640250,196640010,196640251

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.