Learn More
Lenz Laborglasintrumente™ Soxhlet Extractor Head
Extractor heads acc. to Soxhlet
Brand:  Lenz Laborglasintrumente™ 05400143
Product Code. 10284191
  • £164.00 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extractor heads acc. to DIN 12602 and in addition to DIN standard
  • With cone NS 29/32
  • Includes glass NS stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
150 mL
DURAN Tubing
Glass NS stopcock
NS 45/40
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente™ Soxhlet Extractor Head > Capacity (Metric): 150 mL