missing translation for 'onlineSavingsMsg'
Learn More

Wilmad-LabGlass™ SP Scienceware™ Quick Pressure Valve (QPV) 5mm OD NMR Sample Tubes

Low pressure/vacuum (LPV) tube ideal for anaerobic and gas-tight NMR experiments, and offers a convenient flame-free sealing solution for air sensitive or volatile liquid samples

Brand:  Wilmad-LabGlass™ 66306-0575

Product Code. 15318868

  • £181.00 / Each
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Due to the nature of glass, Extreme Caution should be exercised when using at elevated or reduced pressures since a tiny scratch on the glass surface would significantly lower the tensile strength. Adequate safety shielding should always be used when working in these conditions


Includes: Glass tube, piston and vacuum adaptor
This item is not returnable. View return policy

Description

Description

  • Robust sealing system allows pressure build-up inside the sample 
  • Greaseless PTFE piston provides a 100% contamination-free seal
  • Redesigned with a 4X larger sealing surface, eliminating leaks and greatly increasing lifetime when compared to traditional J-Young tubes
  • Axial symmetric design guarantees application in spinning experiments
Specifications

Specifications

6 μm
177.8 mm
5 mm
7 in.
13 μm
Medium Wall Precision Quick Pressure Valve NMR Sample Tube
300 MHz
524-QPV-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Wilmad-LabGlass™ SP Scienceware™ Quick Pressure Valve (QPV) 5mm OD NMR Sample Tubes > Model 535-LPV-7; 600MHz; 7 in. L; 5mm O.D.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.