missing translation for 'onlineSavingsMsg'
Learn More

Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes

Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.

Brand:  Wilmad-LabGlass™ 66999-5700

128.79 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 15309038

  • £198.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
  • WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Specifications

Specifications

60 μm
ASTM E438 Type 1 Class B Glass
Disposable NMR Cap
High-throughput
0.43 mm
WG-1000-7
178 mm
NMR Tube
4.94665±0.01905 mm
7 in.
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    22

    Hours

    6

    Minutes

    17

    Seconds

    26
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721

    Expires2025-05-01

    New Offer
    Save Now
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081

    Expires2025-12-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes > High-Throughput, Length: 178mm, 100/pk

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.