Learn More
Unity Lab Services™ Centrifuge Plus Requalification Service
Requalification Services for Centrifuge Plus (documentation included)
Brand:  Unity Lab Services™ RQCENPLUS
Product Code. 30178676
  • £1724.00 / 1 unit
Explore more special offers
Description
Description
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests Min/Max speed, Brief /Extended time, temperature for refrigerated models.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Centrifuge
Requalification Service
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Unity Lab Services™ Centrifuge Plus Requalification Service > Requalification Service