Learn More
Bel-Art™ Sterileware™ Economy Sample Spoons
For retrieving small, specific volume of powdered or liquid samples
Brand:  Bel-Art™ F36740-0110
 View more versions of this product
Product Code. 15510112
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Made from high-impact polystyrene, FDA grade and sterilized by gamma irradiation
  • Comes with flat bottom shape to allow use as weighing boat
  • Available in white color
  • Available in individual packs for one time use
  • Includes certificate of processing for sterilization
Specifications
Specifications
Spoon
Not Autoclavable
Flat
Single-use
White
Ideal for retrieving small, specific volume samples of powders or liquids, provides durability and stiffness when sampling packed powders
Polystyrene
Certificate of processing for sterilization
1.25 mL
Sterile
FDA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ Sterileware™ Economy Sample Spoons > Bowl capac.: 0.04oz. (1.25mL)