Learn More
Bel-Art™ SP Scienceware™ Sterile Economy Sample Spoons
For retrieving small, specific volume of powdered or liquid samples
Brand:  Bel-Art™ F36740-0210
Code : NEW
Additional Details : Weight : 0.07620kg
 View more versions of this product
Product Code. 15520112
  • £22.75 / Pack of 10
Estimated Shipment: 03-09-2024
to see stock.
Add to basket
Description
Description
  • Made from high-impact polystyrene, FDA grade and sterilized by gamma irradiation
  • Comes with flat bottom shape to allow use as weighing boat
  • Available in white color
  • Available in individual packs for one time use
  • Includes certificate of processing for sterilization
Specifications
Specifications
Spoon
Certificate of processing for sterilization
2.5 mL
Sterile
FDA
Polystyrene
Flat
Yes
White
Ideal for retrieving small, specific volume samples of powders or liquids, provides durability and stiffness when sampling packed powders