missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Spade Tip Stainless Steel Tweezers
Recommended for micro electronics and precision assembly
Brand: Cole-Parmer™ 07288-07
This item is not returnable.
View return policy
Description
- Finely crafted stainless steel tweezers
- Feature hand finished points for accuracy and excellent tip hardness
- Anti-glare finish reduces eye strain
- Anti-magnetic, anti-acid and acid resistant
Specifications
Flat | |
Stainless Steel | |
Tweezers | |
Sterile |
4.5 in. | |
10.07 cm | |
Straight Tweezers |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Spade Tip Stainless Steel Tweezers > Spade Tip Stainless Steel Tweezers
Spot an opportunity for improvement?Share a Content Correction