Learn More
BINDER™ Specimen Temperature Display
Flexible Pt100 indoor temperature sensor
Brand:  BINDER™ 8012-2549
 View more versions of this product
Product Code. 30238262
  • £824.00 / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Platinum 100 ohm temperature sensor for sample temperature display or control.
Specifications
Specifications
KB 65, KB 130, KB 260, KB 470, KB 720, KB 1060, KB 1600
Steel
Temperature Sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Specimen Temperature Display > For use with KB 65, KB 130, KB 260, KB 470, KB 720, KB 1060, KB 1600