Learn More
MP Biomedicals™ SPINeasy™ PCR Purification and Gel Extraction Kit
Spin column kit for quick and convenient DNA clean up from PCR and other enzymatic reactions. Isolate and purify DNA fragments from agarose gels using spin column technology. Purified DNA, free of primers, enzymes, nucleotides, agarose, etc., is ready for downstream molecular biology applications.
Brand: MP Biomedicals™ 116538050
Description
- The SPINeasy PCR Purification and Gel Extraction Kit is a silica -membrane spin column kit that enables quick and convenient DNA clean up from various enzymatic reactions, such as PCR and restriction digestion, as well as isolation and purification of DNA fragments from agarose gels.
- Up to 23 µg of DNA of molecular weight ranging from 100 bp to 20 kb can be purified through a quick and simple process from most grades of agarose gels with expected recovery of 85%.
- Purified DNA is immediately ready for routine molecular biology applications.

Specifications
Store at Room Temperature, Dry Environment | |
50 mL Binding Buffer PG, 6 mL Wash Buffer PG, 10 mL Elution Buffer PG, 50 x Column PG with collection tubes | |
50 |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- SPINeasy PCR Purification and Gel Extraction Kit, 50 preps
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Guanidine Thiocyanate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.