Learn More
Tygon SPT-3350 Platinum-Cured Silicone Tubing
Ultra-smooth inner surface reduces particle entrapment
£102.23 - £4206.72
Specifications
Certifications/Compliance | USP;NSF-51;FDA |
---|---|
Material | Tygon 3350 |
Overall Length (English) | 50 ft. |
Sterilization Method | Yes |
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity & Availability | |||||
30053509
|
Tygon
50114-02 |
1/4 in. | 3/8 in. |
£274.37
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053959
|
Tygon
50114-07 |
3/8 in. | 1/2 in. |
£383.35
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053098
|
Tygon
50113-98 |
3/16 in. | 5/16 in. |
£250.26
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30054135
|
Tygon
50114-00 |
3/16 in. | 7/16 in. |
£493.78
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053030
|
Tygon
50113-90 |
1/16 in. | 1/8 in. |
£115.73
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053420
|
Tygon
50113-92 |
3/32 in. | 5/32 in. |
£125.37
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053915
|
Tygon
50114-10 |
7/16 in. | 9/16 in. |
£438.81
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053674
|
Tygon
50114-09 |
3/8 in. | 5/8 in. |
£710.76
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053763
|
Tygon
50114-01 |
1/4 in. | 5/16 in. |
£190.96
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053277
|
Tygon
50114-17 |
3/4 in. | 1 in. |
£1213.22
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053357
|
Tygon
50114-08 |
3/8 in. | 9/16 in. |
£546.81
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053193
|
Tygon
50114-14 |
1/2 in. | 3/4 in. |
£895.94
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053544
|
Tygon
50114-16 |
5/8 in. | 7/8 in. |
£1072.42
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053721
|
Tygon
50114-18 |
1 in. | 1-1/4 in. |
£1514.11
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30054107
|
Tygon
50114-12 |
1/2 in. | 5/8 in. |
£513.07
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053476
|
Tygon
50114-11 |
7/16 in. | 5/8 in. |
£633.61
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30054184
|
Tygon
50113-99 |
3/16 in. | 3/8 in. |
£357.79
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053526
|
Tygon
50114-13 |
1/2 in. | 11/16 in. |
£678.93
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053711
|
Tygon
50114-20 |
1-1/2 in. | 2 in. |
£4206.72
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053015
|
Tygon
50113-91 |
1/16 in. | 3/16 in. |
£157.69
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30054333
|
Tygon
50113-96 |
5/32 in. | 7/32 in. |
£141.29
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053671
|
Tygon
50113-89 |
1/32 in. | 3/32 in. |
£102.23
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053825
|
Tygon
50114-19 |
1-1/4 in. | 1-1/2 in. |
£1804.40
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30054156
|
Tygon
50113-93 |
3/32 in. | 7/32 in. |
£185.17
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053351
|
Tygon
50114-04 |
1/4 in. | 1/2 in. |
£567.07
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053768
|
Tygon
50113-97 |
3/16 in. | 1/4 in. |
£166.85
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053003
|
Tygon
50114-15 |
5/8 in. | 13/16 in. |
£825.53
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053126
|
Tygon
50114-05 |
5/16 in. | 7/16 in. |
£341.40
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053099
|
Tygon
50114-06 |
5/16 in. | 1/2 in. |
£483.16
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053335
|
Tygon
50114-03 |
1/4 in. | 7/16 in. |
£419.03
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053823
|
Tygon
50113-95 |
1/8 in. | 1/4 in. |
£195.77
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
30053559
|
Tygon
50113-94 |
1/8 in. | 3/16 in. |
£134.53
15m |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
Description
Ideal for food and beverage transfer. Minimal e in., tractable maintains fluid integrity. Up to three times smoother than other silicone tubing. FDA 21 CFR 175.300 compliant. Free technical application support available to assist with tubing selectionSpecifications
USP;NSF-51;FDA | |
Tygon 3350 | |
50 ft. | |
Yes |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Tygon SPT-3350 Platinum-Cured Silicone Tubing