Learn More
Tygon SPT-3350 Platinum-Cured Silicone Tubing
Ultra-smooth inner surface reduces particle entrapment
Brand:  Tygon 50113-93
 View more versions of this product
Product Code. 30054156
  • £185.17 / 15m
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ideal for food and beverage transfer. Minimal e in., tractable maintains fluid integrity. Up to three times smoother than other silicone tubing. FDA 21 CFR 175.300 compliant. Free technical application support available to assist with tubing selection
Specifications
Specifications
USP;NSF-51;FDA
Tygon 3350
50 ft.
Translucent
3/32 in.
7/32 in.
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tygon SPT-3350 Platinum-Cured Silicone Tubing > 3/32 in., 7/32 in.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.