missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Spunbound/Meltblown Bonded Pad

This Spunbound/Meltblown Bonded Pad is a 2-ply, green, medium weight, bonded chemical sorbent pad that absorbs chemicals and hazardous liquids. One sided spunbound gives extra strength as well as less lint. Does not degrade or cause a dangerous chemical reaction with the absorbed liquid.

Brand:  Brady™ 813843

Product Code. 10555685

  • £163.00 / 1 pack
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Green color alerts workers that the product is being used for hazardous liquids.
  • Green color also allows the user to identify and separate hazardous waste for disposal, resulting in lower disposal costs.
  • Perforated for more flexibility - use only what you need and minimize waste.
  • Perfect to clean up chemical spills or for use in laboratories.
Specifications

Specifications

UN1212-E
PAD, Medium Weight, Bonded 30 cm x 30 cm, 50 Pads/Bag, 4 Bags/Disp. Box
80 L
Absorbent Pad
Overspray, leaks, drips, in drum and liquid storage areas, workstations and laboratories.
200 Pk.
11.81 in.
11.811 in.
6.3 kg
Green
20.341 gal.
Chemical Sorbent
Beverage processing, Chemical, Food processing, Laboratory, Manufacturing, Maritime, Oil & gas, Pharmaceutical, Process, Warehouse & logistics
Low-linting
30 x 30 cm
30 cm
30 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Spunbound/Meltblown Bonded Pad > Spunbound/Meltblown Bonded Pad; Dimensions (L x W): 300 x 300 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.