Learn More
AMSTA Rostfritt Stainless Steel Container with Lid
High-quality stainless steel container, available in various sizes with or without clamps and tap.
£361.00 - £3275.00
Specifications
Product Type | Can |
---|---|
Color | Silver |
Material | Stainless Steel |
Finish | Ra 0.2 to 0.3 μm |
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
11741806
|
AMSTA Rostfritt
5060381 |
8 L | Lid |
£361.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11781766
|
AMSTA Rostfritt
506081001 |
100 L | Lid |
£1265.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11781944
|
AMSTA Rostfritt
506081001+5051310X4 |
100 L | Lid and 4 Clamps |
£1510.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11966423
|
AMSTA Rostfritt
506081001+5051310X4+50806 |
100 L | Lid, Tap, and 4 Clamps |
£3275.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11791766
|
AMSTA Rostfritt
506081501 |
150 L | Lid |
£1845.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11791944
|
AMSTA Rostfritt
506081501+5051310X6 |
150 L | Lid and 6 Clamps |
£2210.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11976423
|
AMSTA Rostfritt
506081501+5051310X6+50806 |
150 L | Lid, Tap, and 6 Clamps |
£3155.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11701964
|
AMSTA Rostfritt
506081501+50806 |
150 L | Lid and Tap |
£2575.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11731766
|
AMSTA Rostfritt
50608201 |
20 L | Lid |
£753.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11916423
|
AMSTA Rostfritt
50608201+5051310X3+50806 |
20 L | Lid and 3 Clamps |
£1390.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11701954
|
AMSTA Rostfritt
50608201+50806 |
20 L | Lid and Tap |
£1305.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11741766
|
AMSTA Rostfritt
50608251 |
25 L | Lid |
£628.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11741944
|
AMSTA Rostfritt
50608251+5051310X3 |
25 L | Lid and 3 Clamps |
£1180.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11926423
|
AMSTA Rostfritt
50608251+5051310X3+50806 |
25 L | Lid, Tap, and 3 Clamps |
£1595.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11711954
|
AMSTA Rostfritt
50608251+50806 |
25 L | Lid and Tap |
£1415.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11751766
|
AMSTA Rostfritt
50608301 |
30 L | Lid |
£965.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11936423
|
AMSTA Rostfritt
50608301+5051310X3+50806 |
30 L | Lid, Tap, and 3 Clamps |
£1635.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11721954
|
AMSTA Rostfritt
50608301+50806 |
30 L | Lid and Tap |
£1450.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11761944
|
AMSTA Rostfritt
50608501+5051310X3 |
50 L | Lid and 3 Clamps |
£1015.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11946423
|
AMSTA Rostfritt
50608501+5051310X3+50806 |
50 L | Lid, Tap, and 3 Clamps |
£1780.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11731954
|
AMSTA Rostfritt
50608501+50806 |
50 L | Lid and Tap |
£1610.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Quality
- Invisible weld seams
- Pharmaceutical grade finish
- Sustainable production
- Made in Sweden
Specifications
Can | |
Silver | |
Stainless Steel | |
Ra 0.2 to 0.3 μm |