Learn More
Nickel Electro™ Stainless Steel Beakers with Spout
Will not shatter or chip, this durable stainless steel laboratory beaker is suitabale for laboratory applications.
£102.00 - £293.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11756567
|
Nickel Electro™
201 |
200 mL |
£102.00
Each |
In Stock
Log in to see stock. ![]() |
||||||
12737856
|
Nickel Electro™
202 |
350 mL |
£111.00
Each |
Estimated Shipment: 01-05-2025
Log in to see stock. |
||||||
12747856
|
Nickel Electro™
203 |
550 mL |
£136.24
Each |
In Stock
Log in to see stock. ![]() |
||||||
11766617
|
Nickel Electro™
203 |
550 mL |
£293.00
Pack of 5 |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
11419569
|
Nickel Electro™
204 |
700 mL |
£133.00
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
11429569
|
Nickel Electro™
206 |
1000 mL |
£138.00
Each |
In Stock
Log in to see stock. ![]() |
||||||
11439569
|
Nickel Electro™
205 |
1250 mL |
£139.40
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
11449569
|
Nickel Electro™
207 |
2000 mL |
£233.00
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||||
11459569
|
Nickel Electro™
208 |
2500 mL | N/A | |||||||
11469569
|
Nickel Electro™
209 |
5000 mL |
£227.00
Each |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Durable stainless steel laboratory beaker
- 304 spec stainless steel beakers
- Will not shatter or chip
- Suitable for laboratory applications since
- All surfaces are non-reacting
- Offer zero absorption
Specifications
Stainless Steel | |
Spout | |
Beaker |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Beakers with Spout