missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Stainless Steel Multilevel Slot Sampler
Constructed of robust stainless steel, making it suitable for sampling free flowing powders, granules and even slightly cohesive powders. Sampling Systems™ Stainless Steel Multilevel Slot Sampler is for taking a large volume, cross sectional sample from a container.
Brand: Sampling Systems™ 1210B-1500
This item is not returnable.
View return policy
Description
- Made from high quality 316L stainless steel
- Cross section (Composite sample)
- Suitable for taking large volume samples
Specifications
Slot Sampler | |
5 Slots, Bottle adapter, One bottle | |
25 mm |
Stainless Steel (316L) | |
300 mL | |
1500 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sampling Systems™ Stainless Steel Multilevel Slot Sampler > DiameterMetric: 25mm
Spot an opportunity for improvement?Share a Content Correction