Learn More
Palintest™ Standard Comparator Kit
For use with Palintest™ comparator
Brand:  Palintest™ PT220
Product Code. 11988996
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Palintest™ comparator with interchangeable color discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.
Specifications
Specifications
4 Square test tubes in moulded plastic carrying case with space for 4 Reagent systems Palintest
1 Pack
Standard Comparator Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Palintest™ Standard Comparator Kit > Range: 0 to 100mg/L