missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Starter Set 100 μL 384-format: Eppendorf Xplorer™ Plus Multichannel

From blue sky research to clinical applications, Eppendorf Xplorer / Xplorer plus electronic pipettes provide simplicity, precision, and reproducibility. Available as single-, 8-, 12-, 16-, and 24-channel pipettes, as well as Move It™ adjustable tip spacing variant, for your challenging work.

Brand:  Eppendorf 4861000952

3176.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 30058126

  • £3188.70 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Electronic air-cushion pipette reaching new levels of simplicity, precision and reproducibility
  • Intuitive handling via multi-function rocker with 'up is up' (aspiration) and 'down is down' (dispensing) logic and selection dial to access all pipetting mode
  • Self-explanatory color display without cluttered submenus and with ergonomic viewing angle for optimal readability in every position
  • Spring-loaded tip cone in all Xplorer and Xplorer plus pipettes up to 1,000 μL reduces tip attachment and ejection forces
  • Powerful rechargeable battery for up to 8 hours use & work can continue while charging via power socket
  • Pre-programmed and individual adjustment to external conditions (e.g. geographic height, liquids with high density, elongated tips)
  • Autoclavable lower parts for easy decontamination (all models, incl. Move It™)
  • 9 display languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
  • Xplorer plus only: Individual programming, fixed volume settings, password protection, additional applications, display of service intervals
  • ACT™ - labeled single-channel pipettes and epT.I.P.S.™ pipette tips help you make an informed choice when it comes to sustainability
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lot-Specific Certificate
Autoclavable
Single-use
Sterile
5 to 100 μL
Air-cushion System
19,020 Tips (50 Racks x 384 Tips)
epT.I.P.S.™, epT.I.P.S.™ 384
Light Yellow
16
Electronic Pipettes
Pipetting of aqueous liquids
1 Xplorer™ plus, 16-channel, variable, 5 to 100 μL, light yellow with 5x ep Dualfilter T.I.P.S.™, PCR clean and sterile, 5 to 100 μL, 53 mm, yellow, 19020 tips (50 racks x 384 tips)
0.0416666667
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf Starter Set 100 μL 384-format: Eppendorf Xplorer™ Plus Multichannel > Number of Channels: 16

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.