Learn More
Buerkle™ SteriPlast™ Blue Food Scoops



For sampling powders, granulates and pastes. Buerkle™ SteriPlast™ Food Scoops are sterilized by gamma rays. The color blue does not occur in a natural form in foodstuffs. Scoops can be quickly found and easily identified in a visual check if ever mixed into production.
Brand: Buerkle™ 5378-3013
Description
- Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
- This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
- Can be used as part of HACCP/IFS foreign object management
- Individually packaged for disposable use
- Production, assembly and packaging according to clean room class 7
- Can be detected by colour scanners
Compliance
EU food requirements
Specifications
Food Sampling Scoop | |
Not Autoclavable | |
250 mL | |
Sterile | |
Sampling Powders, Granulates and Pastes | |
232 mm |
Polystyrene | |
8.45 oz. | |
Single-use | |
Blue | |
9.1 in. |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Buerkle SteriPlast Detectable Scoops
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
243852 | Certificate of Compliance | 04/11/2024 | 5378-3013 |
241912 | Certificate of Compliance | 04/07/2024 | 5378-3013 |
233205 | Certificate of Compliance | 08/08/2023 | 5378-3013 |
231404 | Certificate of Compliance | 27/04/2023 | 5378-3013 |
230243 | Certificate of Compliance | 15/02/2023 | 5378-3013 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.