missing translation for 'onlineSavingsMsg'
Learn More

Specac™ Storm™ 10 Short Pathlength Gas Cell FT-IR Stainless Steel 10cm

Brand:  Specac™ GS05800

Encompass_Preferred

Product Code. 15230028

  • £1606.88 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

A 10cm Stainless Steel Gas Cell ideal for analysing gases and vapours at room temperature and low pressures. Samples can be examined in static or flow modes and are demountable with various choices of window materials. Supplied with a slide mounted cell holder for use in an FTIR instrument.
Specifications

Specifications

Gas Cell
10 cm
A slide mounted cell holder for use in an FTIR instrument
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Specac™ Storm™ 10 Short Pathlength Gas Cell FT-IR Stainless Steel 10cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.