Learn More
IBA Lifesciences GmbH Strep-Tactin™ PE for Fab Streptamers™
IBA Lifesciences GmbH Strep-Tactin™ PE for Fab Streptamers™ is for reversible cell isolation delivering label-free target cells.
Brand:  IBA Lifesciences GmbH 6-5001-005
Product Code. 16421485
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Low-affinity Fab-Strep molecules are reversibly multimerized on Strep-Tactin™ PE or APC forming a fluorescent Fab Streptamer™ for cell staining
  • Treatment of labeled target cells with the competing Strep-Tactin™ ligand biotin causes a disruption of the Fab Streptamer™ complex and results in spontaneous dissociation of the Fab-Streps from the target cells
  • The result is a functional and label-free target cell
Specifications
Specifications
All required reagents for cell isolation
For 5 x 107 to 1 x 108 cells
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IBA Lifesciences GmbH Strep-Tactin™ PE for Fab Streptamers™ > Includes: All required reagents for cell isolation