missing translation for 'onlineSavingsMsg'
Learn More

Cytiva StrepTrap XT Chromatography Column

StrepTrap XP is a column packed with Strep-Tactin™ XT Sepharose resin to purify Strep-tag™ II and Twin-Strep-tag™ recombinant proteins.

Brand:  Cytiva 29401317


 View more versions of this product

Product Code. 16522542

  • £216.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For use with Strep-Tactin™ XT Sepharose™
  • Prepacked in chromatography columns for convenience
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
1 x 1 mL
1 mL/min.
Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™
7 mm
25 mm
Affinity Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva StrepTrap XT Chromatography Column > Packaging Size: 1 x 1 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.