missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stuart™ Pressmatic Glass PTFE Ceramic Dispensers

Sets precise volume using a robust, easy to use, self-locking rotating ring. Stuart™ Pressmatic Glass PTFE Ceramic Dispensers deliver priming reagent straight back into the bottle using a recirculation valve to make use safer and less wasteful.

Brand:  Cole-Parmer™ 10112-80

 View more versions of this product

Product Code. 12090892

  • £570.00 / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Pressmatic dispensers have an innovative design that maximizes safety and ease of use. A recirculation valve means that reagent used for priming is delivered straight back into the bottle making it safer and less wasteful. Once the dispenser has been fitted to the bottle, it is fully rotational for optimum positioning. The precise volume setting is by a self-locking rotating ring that is robust, easy to use and will not move during operation. The piston is manufactured from high-grade ceramic eliminating abrasion and sticking. The liquid pathway is made up of glass, PTFE and ceramic to give outstanding chemical resistance. The whole unit can be autoclaved without dismantling. Each Pressmatic dispenser is supplied with 3 x bottle adapters to fit most types of vessel and a works calibrated certificate. A full range of spare parts are available.
  • New ergonomic design with recirculation valve
  • Chemically resistant and non-sticking ceramic piston
  • Simple and accurate volume setting
  • Fully rotational on bottle for optimum positioning
  • Accuracy within 0.7%.
  • Supplied with calibration certificate
  • Can be autoclaved at 121°C
Specifications

Specifications

<0.7%
Autoclavable
0.4 to 2 mL
0.05 mL
Bottletop Dispenser
Glass, PTFE, Ceramic
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Stuart™ Pressmatic Glass PTFE Ceramic Dispensers > Capacity: 0.4 to 2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.