missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Sucrose, Molecular Biology Grade Product Code.: 15446759

Thermo Scientific Chemicals Sucrose, Molecular Biology Grade

Product Code. 15446759
500 g
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15446759

Brand: Thermo Scientific Chemicals J65148.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For protein purification by sucrose density gradients

Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 185°C to 187°C
Color White
Density 1.58
Odor Odorless
Quantity 500 g
Beilstein 90825
Solubility Information Soluble in water
Formula Weight 342.3
Grade Molecular Biology
Physical Form Crystalline Solid
Chemical Name or Material Sucrose
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ18K004 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberJ65148.36,J65148.22,J65148.A1
Lot NumberZ18K005 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberJ65148.36,J65148.22,J65148.A1
Lot NumberZ29J005 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog NumberJ65148.36,J65148.22,J65148.A1
Lot NumberU25J009 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ65148.36,J65148.22,J65148.A1
Lot NumberU25J008 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ65148.36,J65148.22,J65148.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sucrose, Molecular Biology Grade >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.