Learn More
SymLine Tubes, FlexTube & FlexTube Pro
The two most flexible and adaptable tubing systems for the safe disposal of liquid HPLC waste - flexible, expandable, chemically resistant PTFE tubing with a large bending radius.
£22.80 - £391.00
Products 13
Product Code Brand Product Type Diameter (Metric) Outer Overall Length (Metric) Overall Length (English) Price Quantity & Availability  
Product Code Brand Product Type Diameter (Metric) Outer Overall Length (Metric) Overall Length (English) Price Quantity & Availability  
15720189
View Documents
SymLine
106575
FlexTube Pro 16.9 mm 600 mm 23.622 in.
£149.00
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
15530275
View Documents
SymLine
106573
FlexTube Pro 16.9 mm 1.5 m 59.0551 in.
£271.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
15815237
View Documents
SymLine
106572
FlexTube Pro 16.9 mm 2.5 m 98.4252 in.
£391.00
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
15896551
View Documents
SymLine
106574
FlexTube Pro 16.9 mm 1 m 39.3701 in.
£204.00
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
15889657
View Documents
SymLine
106569
FlexTube 19.8 mm 600 mm 23.622 in.
£103.00
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
15705223
View Documents
SymLine
106568
FlexTube 19.8 mm 1 m 39.3701 in.
£115.00
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
15606825
View Documents
SymLine
106567
FlexTube 19.8 mm 1.5 m 59.0551 in.
£129.00
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
15808622
View Documents
SymLine
106566
FlexTube 19.8 mm 25 m 98.4252 in.
£157.00
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
10338169
View Documents
SymLine
108016
Tube 6 mm 1 m 39.3701 in.
£22.80
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
10161160
View Documents
SymLine
108017
Tube 8 mm 1 m 39.3701 in.
£27.85
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
10775051
View Documents
SymLine
108018
Tube 10 mm 1 m 39 3/8 in.
£38.95
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
10156694
View Documents
SymLine
108015
Tube 13 mm 1 m 39.3701 in.
£83.55
1m
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
10785051
View Documents
SymLine
108019
Tube 12 mm 1 m 39.3701 in.
£44.95
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
Description
Description
  • Made of PTFE for best possible chemical resistance
  • Adjustable - easy to install: simply shorten tube as required, then mount the optimized screw connection and sealing
  • Electrostatic conductive < 106 (max) Ω
  • 16.9 mm or 19.8 mm outer diameter
  • Available in 4 different standard lengths of 600, 1200, 1500 and 2500mm.
  • FlexTube with inner wall spiral or or FlexTubePro Inner wall smooth
  • Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    Product Title
    SymLine Tubes, FlexTube & FlexTube Pro