Learn More
Applied Biosystems™ TaqMan™ Array Human MicroRNA A Cards v2.0
TaqMan™ Array Human MicroRNA A Cards v2.0
Brand:  Applied Biosystems™ 4398965
Product Code. 11361453
  • £1496.00 / Pack of 4
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
The TaqMan™ Array Human MicroRNA A Card v2.0 contains 384 TaqMan™ MicroRNA Assays enabling accurate quantitation of 377 human microRNAs in as little as 5 hours. Three TaqMan MicroRNA Assay endogenous controls are included to aid in data normalization and one TaqMan™ MicroRNA Assay not related to human is included as a negative control. Details on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Human Pool A v2.1 is required to run this array. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Human Pool A v2.1 for situations where sensitivity is of the utmost importance and⁄or sample is limiting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Array Human MicroRNA A Card
Human
Dried Down
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Megaplex™, TaqMan™
Standard
384-well Microfluidic Card
4°C
AB 7900 HT 384-Well System
FAM
4 pack
Gene Regulation, Epigenetics
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ TaqMan™ Array Human MicroRNA A Cards v2.0 > 4 pack