Learn More
Thermo Scientific™ KaryoStat+™ Genetic Stability Assay Service
KaryoStat+ Genetic Stability Assay Service is a fast and cost-effective alternative to typical G-banded karyotyping.
Brand:  Thermo Scientific™ A52849
326.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17882606
  • £366.00 / Each
Explore more special offers
Description
Description
  • A fast and cost-effective alternative to typical G-banded karyotyping
  • Leverages the array-based CytoScan HT-CMA 96F Assay Kit (906025), offering whole-genome coverage for accurate detection of chromosomal aberrations in human primary and stem cells
The KaryoStat+ Assay Service offers:
  • Fast data delivery – turnaround time of three weeks or less
  • Convenient sample prep – our assay uses frozen cell pellet, eliminating need for shipping live cells
  • Higher resolution – microarray resolution five times higher than traditional G-banding
  • Exceptional support – a dedicated project manager for frequent project communication
  • Broad cell type compatibility – all non-transformed human cell types accepted
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 9001
3 Weeks
Applied Biosystems™
Pluripotent Stem Cell Researchers
1 sample
Genetic Stability Assay Service
Research Use Only
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ KaryoStat+™ Genetic Stability Assay Service > 1 Sample