Learn More
TBE Sample Buffer, 6X, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J60357.AD
54.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Sample Loading Buffer | |
6X | |
50 mL |
Blue | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P23I023 | Certificate of Analysis | 24/02/2023 | J60357.AC,J60357.AD |
W01I012 | Certificate of Analysis | 19/01/2023 | J60357.AC,J60357.AD |
P17I057 | Certificate of Analysis | 12/04/2022 | J60357.AC,J60357.AD |
Q02E503 | Certificate of Analysis | 08/12/2021 | J60357.AC,J60357.AD |
N21D502 | Certificate of Analysis | 08/12/2021 | J60357.AC,J60357.AD |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.