missing translation for 'onlineSavingsMsg'
Learn More

Technico™ Pocket Refractometer

Brand:  Technico 8145.0100M

 View more versions of this product

Product Code. 10422363

  • £222.00 / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Pocket Refractometer
DIGIT-100 ATC
Salinity
250 g
0 to 100 Salinity
Zero calibration with distilled water between 10 and 30°C
Measuring low salinity concentrations of natural or artificial salt in water, with automatic temperature compensation
55 x 80 x 225 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Datasheets

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Technico™ Pocket Refractometer > Scale Range: 0 to 100% Salinity

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.