Learn More
Fisherbrand™ Temperature Strip
Brand:  Fisherbrand™ FIS08/04A
Encompass
 View more versions of this product
Product Code. 11512513
  • £8.13 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The standard 8 level range covers 40°C to 260°C for general purpose temperature monitoring.
Specifications
Specifications
204°C
166°C to 204°C
5°C
166°C
0.02
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Temperature Strip