missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Test Tube Racks

Customize your test tube rack to fit a varietry of test tube external diameters. Nickel Electro™ Test Tube Racks are non-magnetic stainless steel and supplied flat packed ready for assembly.

Brand:  Nickel Electro™ 6873

 View more versions of this product

Product Code. 11429529

  • £91.85 / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel
  • Non-magnetic
  • Easy assembley
  • Capable of being disassembled for cleaning and sterilization purposes
Specifications

Specifications

Test Tube Rack
Tubes (18 mm)
18
1 Ea.
18 mm
Autoclavable
Stainless Steel
Test Tubes with an External Diameter 18 mm
270 x 138 x 70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel Electro™ Test Tube Racks > Capacity: 18 Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.