missing translation for 'onlineSavingsMsg'
Learn More
Tetra-n-butylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals Product Code.: 11377845

Tetra-n-butylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11377845
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11377845

Brand: Thermo Scientific Alfa Aesar L02809.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.

Solubility
Miscible with organic solvents.

Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1Show MoreShow Less
PubChem CID 2723671
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.99
pH 14.0
Flash Point None
Odor Amine-like
Linear Formula [CH3(CH2)3]4NOH
Refractive Index 1.408
Quantity 250 g
UN Number UN3267
Beilstein 3571228
Sensitivity Air Sensitive
Solubility Information Miscible with organic solvents.
IUPAC Name tetrabutylazanium;hydroxide
Formula Weight 259.48
Concentration or Composition (by Analyte or Components) 40% w/w aq. soln.
Chemical Name or Material Tetra-n-butylammonium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249380 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog NumberL02809.0B,L02809.30,L02809.18
Lot Number10247486 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberL02809.0B,L02809.30,L02809.18
Lot Number10247487 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberL02809.0B,L02809.30,L02809.18
Lot Number10246330 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberL02809.30
Lot Number10199612 Certificate TypeCertificate of Analysis Date09/01/2023 Catalog NumberL02809.0B,L02809.30,L02809.18
compliance-icons compliance-icons
Product Identifier
  • Tetra-n-butylammonium hydroxide, 40% w/w aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : N,N,N-tributyl-1-butanaminium, hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetra-n-butylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.