missing translation for 'onlineSavingsMsg'
Learn More
Tetra-n-hexylammonium perchlorate, Thermo Scientific Chemicals Product Code.: 11397358

Tetra-n-hexylammonium perchlorate, Thermo Scientific Chemicals

Product Code. 11397358
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11397358

Brand: Thermo Scientific Alfa Aesar 030830.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4656-81-9
Molecular Formula C24H52ClNO4
Molecular Weight (g/mol) 454.13
MDL Number MFCD00043185
InChI Key RLCKPGXHGKSGOS-UHFFFAOYSA-M
Synonym tetra-n-hexylammonium perchlorate, tetrahexylammonium perchlorate, tetrahexylazanium perchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate, n,n,n-trihexylhexan-1-aminiumperchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate 1:1, tetra-n-hexyl ammonium perchlorateShow MoreShow Less
PubChem CID 78388
SMILES [O-][Cl](=O)(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Quantity 100 g
UN Number UN1479
IUPAC Name tetrahexylazanium;perchlorate
Formula Weight 454.13
Physical Form Crystalline
Chemical Name or Material Tetra-n-hexylammonium perchlorate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR03K015 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog Number030830.22,030830.14,030830.06
Lot NumberM23K039 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog Number030830.22,030830.14,030830.06
Lot NumberT05J026 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog Number030830,030830.22,030830.14,030830.06
Lot NumberT05J024 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog Number030830,030830.22,030830.14,030830.06
Lot NumberN14I040 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number030830.22,030830.14,030830.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetra-n-hexylammonium perchlorate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.