missing translation for 'onlineSavingsMsg'
Learn More

WTW™ TetraCon™ 325-15 Standard Conductivity Cell

The universally usable conductivity measuring cells by WTW set standard in the conductivity measurement. Their 4 electrode technology makes them insensitive to pollution, uninfluenced by polarization effects and therefore precise and reliable in any conditions.

Brand:  WTW™ 301973

Product Code. 10009780

  • £704.00 / Each
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.475 cm-1
0°C to 90°C
15.3 mm
Epoxy, Graphite
IP68
For conductivity measurements at depths down to 20m
Standard Conductivity Cell
15 m
4-electrode, temperature sensor, 15m Length cable and waterproof plug
TetraCon™ 325-15
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
WTW™ TetraCon™ 325-15 Standard Conductivity Cell > Measuring Range: 1mu S/cm to 2S/cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.