missing translation for 'onlineSavingsMsg'
Learn More
Tetrahydropyran, 99%, Thermo Scientific Chemicals Product Code.: 10700791

Tetrahydropyran, 99%, Thermo Scientific Chemicals

Product Code. 10700791
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10700791

Brand: Thermo Scientific Acros 325881000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-68-7
MDL Number MFCD00006585
InChI Key DHXVGJBLRPWPCS-UHFFFAOYSA-N
Synonym tetrahydropyran, tetrahydro-2h-pyran, oxacyclohexane, pentamethylene oxide, 2h-pyran, tetrahydro, tetrahydropyrane, unii-v06i3ilg6b, 2h-tetrahydropyran, v06i3ilg6b, pyeShow MoreShow Less
PubChem CID 8894
ChEBI CHEBI:46941
IUPAC Name oxane
SMILES C1CCOCC1

Specifications

Melting Point -45.0°C
Color Colorless
Density 0.8800g/mL
Boiling Point 88.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H10O
Refractive Index 1.42
Quantity 100 g
Beilstein 17, 12
Merck Index 15, 9362
Specific Gravity 0.88
Solubility Information Solubility in water: 80g/L (25°C). Other solubilities: miscible with alcohol, ether and many other, organic solvents
Molecular Weight (g/mol) 86.13
Formula Weight 86.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetrahydropyran, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467529 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number325885000,325881000
Lot NumberA0465618 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number325885000,325881000
Lot NumberA0465143 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog Number325885000,325881000
Lot NumberA0458950 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog Number325885000,325881000
Lot NumberA0455498 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number325885000,325881000
compliance-icons compliance-icons
Product Identifier
  • Tetrahydropyran
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydropyran, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.