Learn More
Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min, Thermo Scientific Chemicals
Description
Widely used as a catalyst for palladium-catalyzed coupling reactions including Suzuki coupling, Sonogashira coupling, Heck reaction, Negishi coupling and Stille coupling. Used in isomerization, carbonylation, oxidation, and C-C bond formation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 100°C to 105°C |
Assay Percent Range | 99.8% (metals basis), Pd 9% min |
Quantity | 0.5 g |
Beilstein | 6704828 |
Sensitivity | Air sensitive |
Synonym | tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4 |
Solubility Information | Insoluble in water. Soluble in benzene,ethanol and chloroform |
IUPAC Name | palladium;triphenylphosphane |
Formula Weight | 1155.57 |
Physical Form | Crystalline |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X22K057 | Certificate of Analysis | 05/11/2024 | 010548.02,010548.04,010548.09 |
T19K004 | Certificate of Analysis | 31/07/2024 | 010548.02,010548.04,010548.09 |
Q11K046 | Certificate of Analysis | 11/06/2024 | 010548.02,010548.04,010548.09 |
S28J033 | Certificate of Analysis | 24/10/2023 | 010548.02,010548.04,010548.09 |
S13J033 | Certificate of Analysis | 21/06/2023 | 010548.02,010548.04,010548,010548.09 |
Safety and Handling

- Tetrakis(triphenylphosphine)palladium(0)
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Tetrakis(triphenylphosphine)palladium(0)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.