missing translation for 'onlineSavingsMsg'
Learn More
Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min, Thermo Scientific Chemicals Product Code.: 11376058

Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min, Thermo Scientific Chemicals

Product Code. 11376058
2 g
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy

Product Code. 11376058

Brand: Thermo Scientific Alfa Aesar 010548.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrosilation, isomerization, carbonylation

Widely used as a catalyst for palladium-catalyzed coupling reactions including Suzuki coupling, Sonogashira coupling, Heck reaction, Negishi coupling and Stille coupling. Used in isomerization, carbonylation, oxidation, and C-C bond formation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14221-01-3
Molecular Formula C72H60P4Pd
Molecular Weight (g/mol) 1155.59
MDL Number MFCD00010012
InChI Key NFHFRUOZVGFOOS-UHFFFAOYSA-N
PubChem CID 11979704
SMILES [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 100°C to 105°C
Assay Percent Range 99.8% (metals basis), Pd 9% min.
Quantity 2 g
Beilstein 6704828
Sensitivity Air sensitive
Synonym tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
Solubility Information Insoluble in water. Soluble in benzene,ethanol and chloroform
IUPAC Name palladium;triphenylphosphane
Formula Weight 1155.57
Physical Form Crystalline
Chemical Name or Material Tetrakis(triphenylphosphine)palladium(0)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX22K057 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number010548.02,010548.04,010548.09
Lot NumberT19K004 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number010548.02,010548.04,010548.09
Lot NumberQ11K046 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number010548.02,010548.04,010548.09
Lot NumberS28J033 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number010548.02,010548.04,010548.09
Lot NumberS13J033 Certificate TypeCertificate of Analysis Date21/06/2023 Catalog Number010548.02,010548.04,010548,010548.09
compliance-icons
Product Identifier
  • Tetrakis(triphenylphosphine)palladium(0)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : tetrakis(triphenylphosphine)-palladium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.